n-Pentadecane, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-Pentadecane, Pentadecane

MQ100

76509-25ML 76509-5ML
76509-25MLEA 180 GBP
76509-25ML 76509-5ML
n-Pentadecane, Supelco®
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
Boiling Pt: 271
Melting Pt: 13…15 °C
Density: 0.768 g/cm³ (20 °C)
Flash Pt: 132
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b823a5d81fa846050a5d25e1ac8ce5765eeed7b&vid=4f461a0c4143d26d483347596050e7a1a51b2887&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR