Methyl pelargonate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methyl nonanoate, Nonanoic acid methyl ester, Pelargonic acid methyl ester

M-Clarity™ quality level = MQ100

76368-1ML 76368-5ML
76368-1MLEA 48.1 GBP
76368-1ML 76368-5ML
Methyl pelargonate, Supelco®
Methyl pelargonate
Formula: C₁₀H₂₀O₂
MDL Number: MFCD00009569
CAS Number: 1731-84-6
EINECS: 217-052-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13f28c6427a7137b8dcc3f3867271ab44f177779&vid=084789a5a0d23704f4738211b593d0f2d068aed9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR