Methyl palmitate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

M-Clarity™ quality level = MQ100

76159-1G 76159-5G
76159-1GEA 38.5 GBP
76159-1G 76159-5G
Methyl palmitate, Supelco®
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417
Melting Pt: 29.9
Density: 0.8804 g/cm³ (20 °C)
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e2fce1a4cc76b91e7559abfc791ae541b93477&vid=aa6330e4046171e0dcd7616969ad0cdf4e0ec81d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR