1-Octadecanol, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

74723-5G
74723-5GEA 122 GBP
74723-5G
1-Octadecanol, Supelco®
1-Octadecanol
Formula: H₃C(CH₂)₁₆CH₂OH
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 56…57 °C
Density: 0.805…0.815 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=279decbc0bf3241254c13772e98feeaa0903cd03&vid=35ba575e1e2915cbeef04fe7c0ae6dbea0bbc092&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR