Eppendorf® Protein LoBind Plates
Supplier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030504.100 0030504.119 0030504.208 0030504.216 0030508.106 0030508.203 0030524.101 0030528.107 0030628.306 0030624.300 0030504.305
737-2540EA 673 GBP
737-2540 737-2541 737-2544 737-2545 737-0208 737-0209 737-2564 737-0213 737-0238 737-0163 737-0161
Eppendorf® Protein LoBind Plates
Microplates Deep Well Microplates
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimise the risk of sample interference
  • Lot-certified PCR clean purity grade: Free of human DNA, DNase, RNase and PCR inhibitors
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting
More Product Information