Automated cell imaging system, ImageXpress Pico
Supplier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EXPRESSPICO EXPRESSPICOEC
736-0924EA 0 GBP
736-0924 736-0925
Automated cell imaging system, ImageXpress Pico
Microscopes
The ImageXpress Pico automated cell imaging system is more than a digital microscope, it combines high resolution imaging with powerful analysis and intuitive software. Go from sample to results in minutes and get actionable data from your images faster than ever before.
  • The ImageXpress Pico system offers objectives ranging from 4x to 63x and can operate in colorimetric, brightfield, fluorescence, or digital confocal 2-D 'on the fly' deconvolution imaging modes
  • The new Digital Confocal Option is seamlessly integrated into the ImageXpress Pico’s fluorescent
More Product Information