Microplate readers, Multiskan SkyHigh
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A51119500C A51119600C A51119600DPC A51119700C A51119700DPC
736-0923EA 13520 GBP
736-0923 736-0922 736-0931 LABLA51119700C LABLA51119700DPC
Microplate readers, Multiskan SkyHigh
Microplate Readers
The Multiskan SkyHigh is a UV/Visible microplate spectrophotometer designed to be convenient and easy to use for virtually any photometric research application, especially DNA, RNA and protein analysis. It is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed.
  • Fast, easy measurement - low volume nucleic acid or protein samples
  • Simple to use software - enables experiment set-up within minutes
  • Remote access - manage your data from anywhere via the Cloud, USB, or the Microsoft™ OneDrive™ platform