Cell culture plates, 384-well, CELLSTAR®
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
781091_32 781093_40 781086_32 781182_32 781073_40 781080_32 781098_32 781079_40 781092_40 781091ORG 781090_120 781182_NOV
736-0235EA 1330 GBP
736-0235 738-0053 738-0060 738-0063 392-0311 738-0059 738-0062 738-0058 738-0061 GREI781091ORG GREI781090_120 GREI781182_NOV
Cell culture plates, 384-well, CELLSTAR®
Microplates Cell Culture Plates
PS, cell culture-treated, sterile. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • F-bottom wells with excellent optical properties are ideal for microscopic applications
  • Clear bottom plates have pigmented walls and a transparent thin film bottom, ideal of fluorescence or luminescence detection methods
  • Lid enables gas exchange with minimal evaporation
Colour: Solid bottom plates are available clear, black or white; µClear® (clear) bottom plates are available black or white
Colour: Solid bottomMore Product Information