Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
780270_50 780201_50 786201_80 786261_80 780261_50 780271_50 780215_50 780285_50 780280
736-0133EA 746 GBP
736-0133 736-0130 736-0155 736-0156 736-0132 736-0134 736-0131 736-0135 GREI780280
Storage plates, 96-well, MASTERBLOCK®
Microplates Deep Well Microplates
PP. The 0,5 ml, 1 ml and 2 ml MASTERBLOCK® microplates are ideal for storing samples or for cultivating bacteria or yeasts.
  • Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
  • Uniform external dimensions and tolerances with SBS compatible footprint
  • Elevated well walls allows them to be easily sealed with adhesive films, heat seals or CapMats
  • Alphanumeric well coding
  • Available sterile or non-sterile, and in a choice of colours