Microplates, 96-well
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
650101_100 651101_100 655101_100 655161_100 651161_100 650161_100 655077_40 655075_40 655076_40 655074_40 655094_40 655095_40 655096_40 655097_40 655191_50 651191_400 655944BE
391-3605EA 160 GBP
391-3605 391-3610 391-3615 736-0119 736-0223 391-3311 391-3629 736-0228 736-0229 738-0016 738-0020 738-0021 738-0022 738-0023 738-0168 738-0208 GREI655944BE
Microplates, 96-well
Microplates HTS Microplates
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.