Microplate readers, Varioskan™ LUX
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VLBLATD1 VLBLATD0 VLBLATD2 VLBLA0D2 VLBLA0D1 VLBLA0D0 VLBL0TD2 VLBL0TD1 VLBL0TD0 VLBL00D2 VLBL00D1 VLBL00D0 VLB000D2 VLB000D1 VLB000D0 VL0LATD2 VL0LATD1 VL0LATD0 VL0LA0D2 VL0LA0D1 VL0LA0D0 VL0L0TD2 VL0L0TD1 VL0L0TD0 VL0L00D2 VL0L00D1 VL0000D2 VL0000D1 VL0000D0 VL0L00D0 VLBL0TGD0 VLB000GD0 VLB000GD1 VLB000GD2 VLBL00GD0 VLBL00GD1 VLBL00GD2 VLBL0TGD1 VLBL0TGD2 VLBLA0GD0 VLBLA0GD1 VLBLA0GD2 VLBLATGD0 VLBLATGD1 VLBLATGD2
735-0323EA 0 GBP
735-0323 735-0322 735-0324 735-0321 735-0320 735-0319 735-0318 735-0317 735-0316 735-0315 735-0314 735-0313 735-0312 735-0311 735-0310 735-0309 735-0308 735-0307 735-0306 735-0305 735-0304 735-0303 735-0302 735-0301 735-0300 735-0299 735-0298 735-0297 735-0296 735-0295 LABLVLBL0TGD0 LABLVLB000GD0 LABLVLB000GD1 LABLVLB000GD2 LABLVLBL00GD0 LABLVLBL00GD1 LABLVLBL00GD2 LABLVLBL0TGD1 LABLVLBL0TGD2 LABLVLBLA0GD0 LABLVLBLA0GD1 LABLVLBLA0GD2 LABLVLBLATGD0 LABLVLBLATGD1 LABLVLBLATGD2
Microplate readers, Varioskan™ LUX
Microplate Readers
Varioskan™ LUX comes equipped with a range of measurement technologies including absorbance, fluorescence intensity and FRET as standard, and with optional luminescence, AlphaScreen and time-resolved fluorescence (TRF) modules. The instrument selects the measurement wavelength using filters or monochromators, depending on which is optimal for each measurement technology.
  • Automatic dynamic range selects optimal reading range based on signal intensity
  • Smart safety controls help protect instrument and samples from user error
  • Autocalibration and self-
More Product Information