Tubes, Nunc-Immuno™, Nunc™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
444202 470319
735-0018EA 277 GBP
735-0018 391-8281
Tubes, Nunc-Immuno™, Nunc™
Tubes Immunology Tubes
PS with a choice of MaxiSorp™ (hydrophilic) or PolySorp™ (hydrophobic) surfaces.
  • Nunc StarTubes™ with an increased internal surface, give higher sensitivity and faster assay times
  • Very good optical quality and MaxiSorp™ versions have certified binding homogeneity
  • Compatible with standard equipment
Nunc-Immuno™ tubes are used for solid phase immunoassay methods such as IRMA, ELISA and ILMA methods.