Snap caps for centrifuge tubes, Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
99500-1
734-4245EA 0 GBP
734-4245
Snap caps for centrifuge tubes, Corning®
Closures Test/Sample Tube Closures
HDPE, orange.
These snap caps are manufactured to rigorous standards to ensure a leak proof seal. They are designed to fit PYREX® 5ml disposable glass centrifuge tubes.