VWR®, Single-Use Bioprocessing Bags
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
734-3304EA 974 GBP
734-3304 734-3305 734-3306 734-3307 734-3308 734-3309 734-3310 734-3311 734-3312 734-3313 734-3314 734-3315 734-3316 734-3317 734-3318 734-3319 734-3320
VWR®, Single-Use Bioprocessing Bags
Bags Media Bags
Avantor single-use bioprocessing bags are designed to transfer and store your products throughout the drug manufacturing process. Made from proprietary FlexFilm™ material - a thick, co-extruded film featuring polyethylene inner and outer layers that provide extra resistance to a wide range of chemicals - Avantor single-use bags are supplied in a range of standard sizes to meet the needs of many bioprocessing applications.
  • Bag film made from proprietary ULDPE with high clarity and flexibility
  • Standard 2D bag assemblies provided at volumes ranging from 50 ml
More Product Information