VWR® 3-D scaffold, Cell Culture Dishes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
734-2967EA 476 GBP
734-2967 734-2968 734-2969
VWR® 3-D scaffold, Cell Culture Dishes
Culture Dishes Cell Culture Dishes
VWR® 3-D scaffold cell culture dishes, made of polystyrene (GPPS) with polystyrene polymer scaffold, are able to simulate the three-dimensional structure of cells in animals and the human body to maximum effect, providing an ideal environment for cell interaction, maximising culture area and improving yield.
  • Highly consistent scaffold (mean fibre diameter of 500 with 260 μm pore spacing) with 3-D channels providing high connectivity, facilitating nutrient transmission, consistent metabolic activity, and the accuracy of culture results
  • Cytokine and growth
More Product Information