Counting chambers, Cellometer®
Supplier: NEXCELOM BIOSCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CHT4-SD100-314 CHT4-SD100-514 CHT4-SD100-014 CHT4-SD100-002
NEXCCHT4-SD100-314EA 6820 GBP
NEXCCHT4-SD100-314 734-2902 734-2887 734-2886
Counting chambers, Cellometer®
Slides Counting Chambers
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
  • Two independent enclosed chambers
  • Compatible with all Cellometer® cell counters
  • Cell suspension (20 μl) loaded via a a single channel pipette