Automated cell counters, Cellometer®
Supplier: NEXCELOM BIOSCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CMAUTO2000 CMMINI CMAUTO1000 MMINI-SK150
734-2775EA 24210 GBP
734-2775 734-2776 734-2774 NEXCMMINI-SK150
Automated cell counters, Cellometer®
Cell Counters and Flow Cytometers Automated Cell Counters
The Cellometer® Auto 1000 is designed for mammalian cells to perform density analysis, size measurement, Trypan Blue assays, analysis of clusters and irregular cells. A large touch screen enables the visualisation of cell morphology and selects saved assays with optimised parameters.
  • Compact Cellometer® Mini with a small footprint
  • Powerful software linked to the equipment via USB
  • Brightfield imaging and pattern-recognition software
  • Individual live cells and dead cells stained with Trypan Blue
  • Cell counting, concentration, diameter and viability
  • Calculation
More Product Information