Roller bottles, Nunc™ InVitro™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1060-20 1060-85 1760-20 2160-05 2160-20 4260-22 1060-05 1860-22
734-2393EA 135 GBP
734-2393 734-2394 734-2396 734-2397 734-2694 734-2695 734-2392 734-2395
Roller bottles, Nunc™ InVitro™
Bottles Roller Bottles
PETG, sterile.
  • Quick-action ergonomic closure reduces wrist stain and increases productivity
  • Wide range of sizes with surface areas from 1050 to 4200 cm²
  • Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
  • Easy-to-read graduations for medium fills
  • Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement