Round cover glasses, Nunc Thermanox
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
174950 174969 174977 174985
734-2117EA 476 GBP
734-2117 734-2172 734-2173 734-2118
Round cover glasses, Nunc Thermanox
Cover Glasses
These cover glasses remain flat during normal use.
  • Resistant to common solvents
  • Autofluorescent from 380 to 545 nm
  • Surfaced treatment on one side for optimal cell attachment and growth