Spinner flasks with angled sidearms, Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4500-125 4500-15L 4500-1L 4500-250 4500-36L 4500-3L 4500-500 4500-6L 4500-8L 4502-15L 4502-36L 4502-3L 4502-6L 4502-8L
734-1907EA 691 GBP
734-1907 734-1908 734-1909 734-1910 734-1911 734-1912 734-1913 734-1914 734-1915 734-1916 734-1917 734-1918 734-1919 734-1920
Spinner flasks with angled sidearms, Corning®
Flasks Spinner Flasks
Borosilicate glass with side arms, non sterile.
  • Baffles enhance aeration and agitation of flask contents
  • Impeller design ensures optimal stirring
  • Complete with impeller and caps
  • Side arm design permit easy access of 25 and 50 ml pipettes