Culture slides, Falcon®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
354104 354108 354114 354118
734-0401EA 645 GBP
734-0401 734-0402 734-0088 734-0089
Culture slides, Falcon®
Slides Chamber Slides
Soda-lime glass slide, PS vessel, lid and tool for loosening chamber.
  • Chamber easily and safely removed using the disposable safety removal tool
  • Pressure-sensitive, biocompatible, acrylic-adhesive gasket remains with the vessel after removal, not on the slide, facilitating further processing or placement of coverslips
  • Blue hydrophobic border defines cell culture areas
  • Wells numbered for easy identification
  • Trays designed for incubator use
Falcon® CultureSlides allow cells to be cultured and then analysed on a glass microscope slide. Cells are grown inMore Product Information