Gelatin culture dishes, Corning® BioCoat™
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
354653 356653
734-0252EA 94.2 GBP
734-0252 734-0323
Gelatin culture dishes, Corning® BioCoat™
Culture Dishes Cell Culture Dishes
PS coated with porcine gelatin. Corning® BioCoat™ gelatin cellware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of normal and transfected cell types, including vascular endothelial, muscle, embryonic stem (ES) and F9 teratocarcinoma cells.
  • Gelatin substrate enhances the attachment of a variety of normal and transfected cell types
  • Pre-treatment with gelatin eliminates time consuming preparation, saving time and money
  • Lot-to-lot consistency ensures reproducible
More Product Information