VWR®, Taq Plus 2X master mixes
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
733-2599EA 690 GBP
733-2599 733-2600 733-2597 733-2598
VWR®, Taq Plus 2X master mixes
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq Plus is an optimised format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
  • Ready to use 2X master mix for greater convenience
  • Significantly reduced set-up time
  • Increased specificity compared to Taq DNA polymerase master mix
  • Reduced risk of contamination
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimised buffer system, including dNTPs and magnesium chloride (1,5 mM).