VWR® ExoCleanUp FAST, One-step PCR Clean-up
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
733-2594EA 3140 GBP
733-2594 733-2593 733-2592 733-2849
VWR® ExoCleanUp FAST, One-step PCR Clean-up
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
VWR® ExoCleanUp FAST PCR reagent is a one-step PCR clean-up reagent for optimal sequencing results, consisting of a balanced combination of a heat-labile exonuclease I (HL-ExoI) and a recombinant shrimp alkaline phosphatase (rSAP).
  • Designed to clean-up PCR products in 5 minutes
  • No need for spin columns or magnetic beads
  • Treatment improves downstream applications, such as DNA sequencing and SNP analysis
Treatment of amplified PCR products with this reagent helps to remove residual primers and single-stranded DNA, and inactivates excess dNTPs by dephosphorylation. After enzymatic treatment at 37 °C for a minimum of 2 minutes, this reagent is completely inactivated by heating at 80 °C for a minimum of 3 minutes.