dNTP mix
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AB-0196
733-0548EA 85.1 GBP
733-0548
dNTP mix
Nucleic Acid Reagents Nucleotides
A combined mixture of  dATP, dCTP, dGTP and dTTP containing 5 mM of each dNTP. Applications include PCR, RT-PCR, DNA labelling, and site-directed mutagenesis.