Storage microplates 384-well polypropylene, Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3656 3657 3964 3965 3658 3342 3347
732-5528EA 533 GBP
732-5528 732-5529 732-5550 732-5551 732-5530 732-0483 736-0202
Storage microplates 384-well polypropylene, Corning®
Microplates Deep Well Microplates
These 384-well polypropylene plates and blocks are built to the industry standard footprint.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks areMore Product Information