Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10426880 10426890 10426892 10427818 10427826 10427810 10427812 10427813 10427804 10427806 10547922 10427805
732-4091EA 31.4 GBP
732-4091 732-4092 732-4093 732-4098 732-4100 732-2531 732-4229 732-4230 732-2760 732-2599 732-2762 732-2761
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Blotting Papers
This general purpose blotting paper is made from pure raw materials with a high absorbency and is used as a membrane gel support. Due to its thickness (0,8 mm), it is recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic and blotting techniques ensuring the wicking capability and uniformity of capillary action
More Product Information