VWR® XT⁹⁶ XTender⁹⁶, Thermal Cyclers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
732-1359EA 4090 GBP
732-1359 732-3428 732-3482 732-3658 732-3657
VWR® XT⁹⁶ XTender⁹⁶, Thermal Cyclers
PCR Cyclers
The XT family of VWR® PCR thermal cyclers combine industry- and customer-inspired innovation housed in a robust, low noise design that fits on virtually any bench top. The fast ramping 96-well block is accurately controlled by eight Peltier elements ensuring reproducible PCR results and offering a gradient PCR option. Making use of advanced technology, XT cyclers can be connected via Ethernet and controlled remotely by a primary XT⁹⁶ cycler, a PC or a mobile device (smartphone/tablet). The software/App needed comes free and provides the same intuitive user interface as the touch-sensitive,More Product Information