PCR plates, 96-well, non skirted, low profile, Thermo-Fast®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AB-0700 AB-0700/W AB-0700/R AB-0700/G AB-0700/B AB-0700/P AB-0700/Y AB-0700-L
732-4851EA 170 GBP
732-4851 732-4853 732-5013 732-5011 732-5010 732-5012 732-5014 732-2943
PCR plates, 96-well, non skirted, low profile, Thermo-Fast®
Microplates PCR Plates
PP. The low profile of the plate decreases ‘dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side-wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
  • Maximum well volume 0,2 ml when used with adhesive and heat seals
  • Cut corner (H12)
  • Certified free from DNase, RNase and human genomic DNA