SE600 Standard Dual Cooled Vertical Electrophoresis Unit

Supplier: HOEFER ELECTROPHORESIS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SE600-15-1.5 SE600
732-1919EA 1780 GBP
732-1919 732-1918
SE600 Standard Dual Cooled Vertical Electrophoresis Unit
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The SE600 is ideal for all standard electrophoresis separations.

  • Maximum glass plate size 18×16 cm
  • Produce straight lanes and sharp, well defined bands
  • Run up to four gels (maximum 112 samples) at one time under identical conditions
  • Run gels at uniform temperature from 1 to 45 °C
  • Accommodates denaturing and native polyacrylamide gels and 2-D electrophoresis
  • A wide array of accessories allows gel configurations to be tailored to individual needs

Delivery information: Complete system includes lower buffer chamber, upper buffer chamber, safety lid withMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR