PCR plates, 96-well, SuperPlate™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AB-2400 AB-2400/W AB-2800 AB-2800/W
732-1060EA 185 GBP
732-1060 732-1063 732-1049 732-1052
PCR plates, 96-well, SuperPlate™
Microplates PCR Plates
PP. Thermo Scientific SuperPlate™ provides four times more rigidity for superior robotic handling. Three versions are available, all with ultra-rigid SBS footprint for compatibility with automated systems, and thin walled round bottom wells for optimal heat transfer and maximum sample recovery.
  • Enhanced thermal stability to minimise warping during thermal cycling and heat sealing
  • Black lettering for quick referencing of wells
  • Maximum well volume 0,2 ml when used with adhesive heat seals
  • Skirted version is stackable and compatible with automated systems
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable