Electrodes, 2-Needle Array™
Supplier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45-0205 45-0120 45-0206 45-0121
732-0029EA 343 GBP
732-0029 732-0018 732-0030 732-0019
Electrodes, 2-Needle Array™
Electrodes Electroporation Electrodes
2-Needle Array™ electrodes are needle-style electrodes specifically designed for in vivo drug or gene delivery applications. The electrode consists of a reusable 2-Needle Array™ handle and sterile, disposable, 2-needle arrays.
  • Stainless steel electrode, delrin handle
  • Simple, inexpensive and safe non viral technique for in vivo gene transfer by the direct injection of plasmid DNA into muscle
The electrode is available in two sizes. The 5 mm 2-needle array and handle is recommended for small muscle masses such as mouse tibialis. The 10 mm 2-More Product Information