Hybridisation apparatus, Big Shot III™ and Little Shot III™
Supplier: BOEKEL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
230402-EU 230402-UK 230402-CH 230502-2-CH 230502-2-EU
731-0512EA 4620 GBP
731-0512 731-0513 731-0511 BOEI230502-2-CH BOEI230502-2-EU
Hybridisation apparatus, Big Shot III™ and Little Shot III™
Ovens Hybridisation Ovens
Hybridisation and wash methods vary according to application and personal choice. Boekel Scientific offers a range of hybridisation incubators with features that include digital display for ease and reproducibility of temperature setting; microprocessor or proportional temperature control; rapid heat-up, optimised heater power and natural or forced air convection.
  • Forced air convection, coupled with PID temperature control, combine to provide rapid heat up and excellent temperature stability
  • Unit is stackable and provided with a reversible door to optimise
More Product Information