Film, Kodak BioMax® MS
Supplier: CARESTREAM HEALTH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1764505 1777853
730-3223EA 0 GBP
730-3223 730-3224
Film, Kodak BioMax® MS
Autoradiography Films Films
BioMax® MS film is a high performance all-purpose, maximum sensitivity autoradiography film. When used with the BioMax® TranScreen HE intensifying screen it delivers four to eight times greater sensitivity in the detection of ³²P when compared to other autoradiography film and intensifying screen combinations. This film/screen combination is ideal for detection of Northern, Southern and Dot Blotting applications, particularly when detection of low intensity samples or exposure time reduction is required.
  • General purpose autoradiography film
  • Particularly recommended for ³²P-labelled
More Product Information