Film for chemiluminescent imaging, Kodak BioMax® Light
Supplier: CARESTREAM HEALTH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1538966 1917012 8194540 8761520
730-3211EA 0 GBP
730-3211 730-3226 730-3227 730-3244
Film for chemiluminescent imaging, Kodak BioMax® Light
Autoradiography Films Films
BioMax® Light is a double emulsion film that provides maximum clarity and sensitivity for detection of chemiluminescent-labelled samples. BioMax® Light produces the highest signal-to-noise ratio of any Kodak film for detection of chemiluminescence. This film performs well with all brands of chemiluminescent kits. BioMax® Light film is suitable for direct and intensifying screen exposures with commonly used isotopes, though it provides lower sensitivity resulting in longer exposure times compared to BioMax® MS, and X-OMAT AR films for radioisotope detection.
  • Suitable for both manual and
More Product Information