Power supplies, EV2xx series

Supplier: CONSORT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EV3020 EV262 EV3020-CH EV3020-UK EV2310-CH EV2310-UK EV2650-CH EV2650-UK
730-1113EA 1020 GBP
730-1113 730-1136 730-1114 730-1115 730-1123 730-1124 730-1140 730-1141
Power supplies, EV2xx series
Power Supplies
A complete range of power supplies for electrophoresis applications, including DNA sequencing, flat bed iso-electric focussing, horizontal and vertical gels, electro-elution, Western blotting and semi-dry blotting.

  • All models are method programmable with up to 9 different programmes each with 9 steps
  • Timer or volt-hour controlled operation: The microcomputer will automatically terminate the run and sound an alarm when the countdown to the selected value is complete
  • Each model can constantly monitor the behaviour of all parameters to prevent depletion of
More Product Information
Order Now

SPECIFICATIONS

Model EV202 EV262 EV202 EV202 EV231 EV231 EV265 EV265
Environmentally Preferable
Voltage 0 - 300 V 0 - 6000 V 0 - 300 V 0 - 300 V 0 - 300 V 0 - 300 V 0 - 600 V 0 - 600 V
Volt-hours 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh 0 - 99,99 kVh
Current 0 - 2000 mA 0 - 150 mA 0 - 2000 mA 0 - 2000 mA 0 - 1000 mA 0 - 1000 mA 0 - 500 mA 0 - 500 mA
Power 0 - 300 W 0 - 300 W 0 - 300 W 0 - 300 W 0 - 150 W 0 - 150 W 0 - 150 W 0 - 150 W
Power requirements 210 - 250 V AC; 50/60 Hz; 360 W 210 - 250 V AC; 50/60 Hz; 360 W 210 - 250 V AC; 50/60 Hz; 360 W 210 - 250 V AC; 50/60 Hz; 360 W 210 - 250 V AC; 50/60 Hz; 200 W 210 - 250 V AC; 50/60 Hz; 200 W 210 - 250 V AC; 50/60 Hz; 200 W 210 - 250 V AC; 50/60 Hz; 200 W
Timer 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h
Display LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters LCD, 2×16 characters
Programs 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters 9×9 set of parameters
Parameter range (full scale) 1 - 100% 1 - 100% 1 - 100% 1 - 100% 1 - 100% 1 - 100% 1 - 100% 1 - 100%
Resolution 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W 1 V;1 mA;1 W
Buffer depletion detection On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable
Min. load resistance 5 (Ω) 1200 (Ω) 5 (Ω) 5 (Ω) 30 (Ω) 30 (Ω) 30 (Ω) 30 (Ω)
Overload detection Yes Yes Yes Yes Yes Yes Yes Yes
Ground leakage detection Yes Yes Yes Yes Yes Yes Yes Yes
Ambient temperature 0...40 °C 0...40 °C 0...40 °C 0...40 °C 0...40 °C 0...40 °C 0...40 °C 0...40 °C
Relative humidity 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing 0 - 95 %, non-condensing
W×D×H 310×260×150 mm 310×260×150 mm 310×260×150 mm 310×260×150 mm 310×260×150 mm 310×260×150 mm 310×260×150 mm 310×260×150 mm
Load detection On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable On/off, programmable
Output 4 in parallel, 4 mm sockets 4 in parallel, 2 mm sockets 4 in parallel, 4 mm sockets 4 in parallel, 4 mm sockets 4 in parallel, 4 mm sockets 4 in parallel, 4 mm sockets 4 in parallel, 4 mm sockets 4 in parallel, 4 mm sockets
Timer 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h 0 - 99:59 h
Weight 10 kg 10 kg 10 kg 10 kg 6 kg 6 kg 6 kg 6 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR