Speci®-Ceps, Heated forceps
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
720-2750EA 0 GBP
720-2750 ITGEHF10 720-2752 720-2753
Speci®-Ceps, Heated forceps
Forceps
Speci®-Ceps are electrically-heated forceps designed for the preparation of histological or histopathological slides.
  • Continuous use, without adjustment, and without influencing the temperature of the medium or tissue
  • Eliminates the need to keep forceps at a usable temperature, enabling the operator to concentrate on orientation of the tissue
  • Results in a significant increase in productivity
To prevent the cold tips of normal forceps from solidifying the liquid wax medium, it is necessary to repeatedly remove the forceps from the work and heat them byMore Product Information