IV catheter with injection port, Vasofix® Braunüle®
Supplier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4268210B 4268130B 4268091B 4268113B
720-2549EA 91.4 GBP
720-2549 720-2548 720-2546 720-2547
IV catheter with injection port, Vasofix® Braunüle®
Catheters
IV catheter material available in PU and FEP for low-pain and gentle insertion. Medication can be quickly administered without having to repuncture the area.
  • Four fully integrated x-ray contrast lines
  • Hydrophobic blood stoppers, removable lock cones
  • Colour-coded sizes
  • Free of latex and PVC
  • Excellent puncture properties with atraumatic tip and triple facet bevel geometry
Same product properties as the Vasofix® Safety (with FEP catheter) excluding the integrated, self-activating safety device, for preventing needlestick injuries and the associated riskMore Product Information