VWR® Q Path® MacroStar II, Cassettes, in Dispenser
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
720-2391EA 226 GBP
720-2391 720-2392 720-2393 720-2394 720-2395 720-2396 720-2397 720-2398 720-2399
VWR® Q Path® MacroStar II, Cassettes, in Dispenser
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with tape (= rack) for Leica and Sakura cassette printers. Rack of 40 cassettes.
  • Large opening for the filtration of fluids
  • Hinge and separate lid
  • 45° writing angle
Order Now
SPECIFICATIONS
Environmentally Preferable