VWR® Q Path® MacroStar II, Cassettes, in Sleeve
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
720-2052EA 416 GBP
720-2052 720-2054 720-2055 720-2056 720-2057 720-2058 720-2051 720-2053 720-2050
VWR® Q Path® MacroStar II, Cassettes, in Sleeve
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes in pre-filled sleeves for Thermo and RA Lamb cassette printers. 50 dispensers with 80 cassettes and 2×2000 lids.
  • Large opening for the filtration of fluids
  • Hinge and separate lid
  • With 45° writing angle