Tissue processing cassette, System II Hex
Supplier: CELLPATH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EAD-0102-03A EAD-0104-03A EAD-0105-03A EAD-0106-03A EAD-0107-03A EAD-0108-03A EAD-0109-03A
720-1891EA 52.5 GBP
720-1891 720-1892 CLLPEAD-0105-03A 720-1893 720-1894 720-1895 720-1896
Tissue processing cassette, System II Hex
Embedding Cassettes Tissue Processing Cassettes
The System II Hex Cassette differs from conventional round hole processing cassettes in that it has hexagonal instead of round perforations arranged in a honeycombed structure to better utilise the available surface area.
  • Available in different stock colours, which allow laboratories to colour code different types of workload or different applications
  • Manufactured from high–grade virgin acetyl resin
This unique arrangement increases the open area by approximately 13 percent thereby increasing reagent flow through and improving specimen staining andMore Product Information