Staining jars according to Columbia, WHEATON®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W900180 W900180-6
720-1035EA 76.4 GBP
720-1035 720-1036
Staining jars according to Columbia, WHEATON®
Jars Staining Jars
Soda-lime glass.
  • Unit holds four cover slips
  • PP screw cap with PTFE-coated PE liner
  • Chemically resistant
These staining jars hold up to four cover slips 17 to 23 mm wide and up to 30 mm long. Longer cover slips can be accommodated if the cap is removed. In addition to staining applications, Columbia jars can be used for cleaning cover slips, as well as coating them with materials such as polylysine or silane.