Slide holders, SlideFolder™
Supplier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M750-20AS M750-20B M750-20G M750-20GY M750-20W M750-20Y M752-20WOP
720-0479EA 288 GBP
720-0479 720-0480 720-0481 720-0482 720-0483 720-0484 630-1028
Slide holders, SlideFolder™
Slide Holders
The SlideFolder is manufactured from heat-resistant, injection moulded plastic and can hold up to 20 standard microscope slides.
  • Numbered spaces and transparent leaves for easier identification
  • Five colours allowing colour coded classification of the slides
  • Slides are removed simply by pressing on one end
  • Can be stacked to save space