Pathology knife, Black Diamond™
Supplier: CELLPATH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CAA-0401-01A CAA-0601-01A CAA-0801-01A CAA-1001-01A CAA-1201-01A CAA-9901-01A
720-0010EA 122 GBP
720-0010 720-0011 720-0012 720-0013 720-0014 720-0015
Pathology knife, Black Diamond™
Knives
Disposable cartilage knife with Sheffield stainless steel double honed blade and molded handle with finger guard.
  • Excellent alternative to resharpening autopsy knives
  • Complete disposal of knife and handle eliminates problems of replacement blade fit and ensures contamination-free environment
  • Available in five different types and lengths (cartilage, trimming, macro, lung and brain knives) to enable pathologist to easily select optimum size of knife for any purpose
Delivery information: Supplied in packs of 10 or an assorted pack containing two knivesMore Product Information