J.T.Baker® BAKERBOND®, Wide-Pore Prep Packings
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7191-00EA 0 GBP
7191-00 7191-02 7191-05 7183-02 7190-02 7190-05 7207-00
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packings
Chromatography Sorbents HPLC Sorbents
Silica-based column packings with high performance purification of strongly hydrophilic peptides and proteins, based on hydrophobic characteristics and molecular shapes.