J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7182-02EA 0 GBP
7182-02 7182-05 7285-00 7285-01
J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Protein Purification Protein Purification Resins
Silica-based media used for the purification of proteins based on differential salt induced hydrophobicity. Mixed mode provides unique selectivity with its hydrophobic interaction and ion exchange function.