Distillation apparatus, Behrotest® WE 5 and associated glassware
Supplier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B00217814 B00218122 B00218197 B00218204 B00218205 B00218206 B00218170 B00218187 B00218124 B00218112 B00218109
BEHRB00217814EA 1180 GBP
BEHRB00217814 718-1251 BEHRB00218197 718-1091 718-1092 718-1093 718-1214 718-1090 718-1252 718-1011 718-1001
Distillation apparatus, Behrotest® WE 5 and associated glassware
Distillation Systems Automatic Distillation Systems
The behrotest® WE 5 Five-in-a-line multiple steam generator provides the simultaneously rapid and gentle distillation of up to five samples. The discharge of steam into the samples guarantees a particularly uniform distillation and thus a high reproducibility of the recovery rates over the entire range. The complete set includes:
  • Temperature and time controller
  • Heating block with five cavities
  • Thermal insulation cover
  • Stand for DET 5 glassware
  • Coil cooler
For the determination of ammonium-nitrogen as defined in DIN 38406-E5 and DIN 38409-H16-2 phenol index procedure.