VWR® SAS Super ISO Air Samplers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710-2087EA 6190 GBP
710-2087 710-2088 710-2089 710-2090
VWR® SAS Super ISO Air Samplers
Samplers Air Samplers
SAS Super ISO 100 and SAS Super ISO 180 are differentiated by their airflow; this choice depends on the application, and allows continuous or sequential sampling from a few minutes to several hours. The data can be stored on the SAS instrument itself or on any remote PC.
  • Design avoids turbulence in unidirectional airflow and re-aspiration of tested air
  • Portable and easily positioned, they operate with long life, rechargeable batteries
  • All manuals are available on a USB key in six languages
  • Micro USB for data download and instrument configuration
  • New Data
More Product Information