VWR® Premium 150, Premium 90 HR, Automatic Colony Counters
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710-2073EA 13250 GBP
710-2073 710-2074
VWR® Premium 150, Premium 90 HR, Automatic Colony Counters
Colony Counters
Premium 150 and Premium 90 HR colony counters and zone measurements systems.
  • Plates can be automatically read at the press of a button
  • High definition colour images taken with a 1,4 MP scientific grade CCD camera ensure that even the smallest colony can be seen and counted, while zone measurements are fast, accurate and reproducible
  • A unique 3-channel LED lighting system configured for exceptional illumination of all sample types
  • Can be used with a new or existing stand-alone desktop or laptop computers via USB connection – no additional PCI cards are
More Product Information