VWR® SAS Super Pinocchio II Air Sampler for Compressed Gases
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710-0950EA 4570 GBP
710-0950 710-0949 710-0955 710-0951
VWR® SAS Super Pinocchio II Air Sampler for Compressed Gases
Samplers Air Samplers
SAS Super Pinocchio II is a microbial impaction sampler to test the quality of compressed air and gases used in cleanrooms. A compressed air source is connected and the flow meter regulated to achieve the required flow rate.
  • Autoclavable
  • Calibrated according to International Standards
  • Unit requires no power and is fully transportable
  • Can collect air samples on either contact plates or 90 mm petri dishes
Certifications: IQ, OQ and PQ documentation available.
Delivery information: Self-contained unit, consisting of connections, airflow meter,More Product Information